Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetraethylene glycol diacrylate ≥90.0% stabilized

Supplier: Decon Laboratories Ltd

T1569-25G T1569-500G
TCT1569-025GEA 29.49 USD InStock
TCT1569-025G TCT1569-500G
Tetraethylene glycol diacrylate ≥90.0% stabilized
Tetraethylene glycol diacrylate
(stabilized with MEHQ) CAS Number: 17831-71-9 MDL Number: MFCD00008630 Molecular Formula: C14H22O7 Molecular Weight: 302.32 Purity/Analysis Method: 90.0% (GC) Form: Clear Liquid Color: Colorless Flash Point (°C): 185 Specific Gravity (20/20): 1.12


Formula: C₁₄H₂₂O₇
Density: 1.12 (20 °C)
Flash Pt: 185 °C
MDL Number: MFCD00008630
CAS Number: 17831-71-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e52383778eac95ce9b3fbac422d122db331a1321&vid=e52383778eac95ce9b3fbac422d122db331a1321&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 90.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty