Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethylsulfonium hydroxide 0.2 mol/L in methanol

Supplier: Decon Laboratories Ltd

T1576-25ML T1576-5ML
TCT1576-025MLEA 360.67 USD InStock
TCT1576-025ML TCT1576-5ML
Trimethylsulfonium hydroxide 0.2 mol/L in methanol
Trimethylsulfonium Hydroxide
[Methylating Reagent]
CAS Number: 17287-03-5
MDL Number: MFCD00216756
Molecular Formula: C3H10OS
Molecular Weight: 94.17
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 11
Specific Gravity (20/20): 0.81
Storage Temperature: <0°C


Formula: C₃H₁₀OS
Density: 0.81 (20 °C)
Flash Pt: 11 °C
MDL Number: MFCD00216756
CAS Number: 17287-03-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f5ca8a1618c392e2503dfed364a842e76f893a2&vid=2f5ca8a1618c392e2503dfed364a842e76f893a2&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty