Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

O-(Trimethylsilyl)hydroxylamine ≥95.0%

Supplier: Decon Laboratories Ltd

T1669-5G T1669-25G
TCT1669-005GEA 198 USD InStock
TCT1669-005G TCT1669-25G
O-(Trimethylsilyl)hydroxylamine ≥95.0%
O-(Trimethylsilyl)hydroxylamine
CAS Number: 22737-36-6 MDL Number: MFCD00010872 Molecular Formula: C3H11NOSi Molecular Weight: 105.21 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 105 Flash Point (°C): 12 Specific Gravity (20/20): 0.87 Storage Temperature: 0-10°C


Formula: C₃H₁₁NOSi
Boiling Pt: 105 °C
Density: 0.87 (20 °C)
Flash Pt: 12 °C
MDL Number: MFCD00010872
CAS Number: 22737-36-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da4515fb360bc24424af94ed3c79ca2bc5fc75d2&vid=da4515fb360bc24424af94ed3c79ca2bc5fc75d2&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (GC)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty