Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trimethylstannyl)pyridine ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

T1742-1G T1742-5G
TCT1742-001GEA 202 USD InStock
TCT1742-001G TCT1742-5G
2-(Trimethylstannyl)pyridine ≥98.0% (by titrimetric analysis)
2-(Trimethylstannyl)pyridine
CAS Number: 13737-05-8
MDL Number: MFCD01631312
Molecular Formula: C8H13NSn
Molecular Weight: 241.91
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.40
Storage Temperature: 0-10°C

Other Product Names / Synonyms: 2-Trimethylstannylpyridine


Formula: C₈H₁₃NSn
Density: 1.40 (20 °C)
MDL Number: MFCD01631312
CAS Number: 13737-05-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89e1d4e9f2b601109be8ab0df9fada9dab02fe55&vid=89e1d4e9f2b601109be8ab0df9fada9dab02fe55&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty