Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetraethylphosphonium tetrafluoroborate ≥97.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

T1747-5G T1747-25G
TCT1747-005GEA 181 USD InStock
TCT1747-005G TCT1747-25G
Tetraethylphosphonium tetrafluoroborate ≥97.0% (by titrimetric analysis)
Tetraethylphosphonium tetrafluoroborate
CAS Number: 665-49-6
MDL Number: MFCD01631314
Molecular Formula: C8H20BF4P
Molecular Weight: 234.03
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White


Formula: C₈H₂₀BF₄P
MDL Number: MFCD01631314
CAS Number: 665-49-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=458b17b91499708eafe93cd6809fab25cea5802e&vid=458b17b91499708eafe93cd6809fab25cea5802e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty