Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Tryptophanamide hydrochloride ≥98.0%

Supplier: Decon Laboratories Ltd

T2312-1G T2312-5G
TCT2312-1GEA 68 USD InStock
TCT2312-1G TCT2312-5G
L-Tryptophanamide hydrochloride ≥98.0%
L-Tryptophanamide hydrochloride
CAS Number: 5022-65-1 MDL Number: MFCD00054315 Molecular Formula: C11H13N3O Molecular Weight: 239.70 Purity/Analysis Method: 98.0% (HPLC,N) Form: Crystal Color: White Melting point (°C): 250 Specific rotation [a]20/D: 25 deg (C=1, H2O)


Formula: C₁₁H₁₃N₃O
Melting Pt: 250 °C
MDL Number: MFCD00054315
CAS Number: 5022-65-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=505fb8d2e8d8830b5b010af7bd7ad016ff74f967&vid=505fb8d2e8d8830b5b010af7bd7ad016ff74f967&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,N)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty