Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(4-nitrophenyl)amine ≥95.0% (by HPLC)

Supplier: Decon Laboratories Ltd

T2333-25G T2333-5G
TCT2333-25GEA 272 USD InStock
TCT2333-25G TCT2333-5G
Tris(4-nitrophenyl)amine ≥95.0% (by HPLC)
Tris(4-nitrophenyl)amine
CAS Number: 20440-93-1 MDL Number: MFCD00024636 Molecular Formula: C18H12N4O6 Molecular Weight: 380.32 Purity/Analysis Method: 95.0% (HPLC) Form: Crystal Color: Yellow Melting point (°C): 280


Formula: C₁₈H₁₂N₄O₆
Melting Pt: 280 °C
MDL Number: MFCD00024636
CAS Number: 20440-93-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f8065f0673f0c868ac2ecd832078694ac926bc9&vid=6f8065f0673f0c868ac2ecd832078694ac926bc9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty