A5551-1G
A5551-5G
TCA5551-001GEA
92
USD
InStock
TCA5551-001G
TCA5551-005G
Br-Mmc (=4-bromomethyl-7-methoxycoumarin) ≥98.0% (by titrimetric analysis)
Br-Mmc (=4-bromomethyl-7-methoxycoumarin)
[for HPLC Labeling]
CAS Number: 35231-44-8
MDL Number: MFCD00006869
Molecular Formula: C11H9BrO3
Molecular Weight: 269.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 215
Storage Temperature: 0-10°C
Product Categories: Analytical Science, Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 4-Bromomethyl-7-methoxycoumarin