Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trichostatin A ≥98.0%

Supplier: Decon Laboratories Ltd

T2477-10MG T2477-50MG
TCT2477-10MGEA 726.89 USD InStock
TCT2477-10MG TCT2477-50MG
Trichostatin A ≥98.0%
Trichostatin A
CAS Number: 58880-19-6
MDL Number: MFCD03848392
Molecular Formula: C17H22N2O3
Molecular Weight: 302.37
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 144
Specific rotation [a]20/D: 136 deg (C=0.3, MeOH)
Storage Temperature: <0°C

Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Regenerative Medicine Research Reagents, Antitumor Ingredients, Epigenetics Research Reagents
Other Product Names / Synonyms: TSA, [R-(E,E)]-7-[4-(Dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxo-2,4-heptadienamide


Formula: C₁₇H₂₂N₂O₃
MW: 302.37 g/mol
Melting Pt: 144 °C
MDL Number: MFCD03848392
CAS Number: 58880-19-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b08c515bba02f7741043c49b7a82897df939952&vid=3b08c515bba02f7741043c49b7a82897df939952&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty