Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethyl(vinyloxy)silane ≥95.0%

Supplier: Decon Laboratories Ltd

T2561-1G T2561-5G
TCT2561-1GEA 33 USD InStock
TCT2561-1G TCT2561-5G
Trimethyl(vinyloxy)silane ≥95.0%
Trimethyl(vinyloxy)silane
CAS Number: 6213-94-1 MDL Number: MFCD00054764 Molecular Formula: C5H12OSi Molecular Weight: 116.24 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 75 Flash Point (°C): -19 Specific Gravity (20/20): 0.78 Storage Temperature: 0-10°C


Formula: C₅H₁₂OSi
MW: 116.24 g/mol
Boiling Pt: 75 °C
Density: 0.78 (20 °C)
Flash Pt: –19 °C
MDL Number: MFCD00054764
CAS Number: 6213-94-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94264270afb70a04bb2923b13a7a55c9eb10c549&vid=94264270afb70a04bb2923b13a7a55c9eb10c549&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty