Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(aminoguanidinium)sulphate ≥98.0% (by titrimetric analysis) for synthesis

Supplier: Decon Laboratories Ltd

A0309-25G A0309-500G
TCA0309-025GEA 30 USD InStock
TCA0309-025G TCA0309-500G
Bis(aminoguanidinium)sulphate ≥98.0% (by titrimetric analysis) for synthesis
Bis(aminoguanidinium)sulphate
CAS Number: 996-19-0
MDL Number: MFCD00035665
Molecular Formula: CH6N4
Molecular Weight: 246.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 207


Formula: C₂H₁₄N₈O₄S
Melting Pt: 207 °C
MDL Number: MFCD00035665
CAS Number: 996-19-0
EINECS: 213-628-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bedd43f0b280c44ed2260e84f2be069ad3ae9be&vid=9bedd43f0b280c44ed2260e84f2be069ad3ae9be&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty