Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Undecanedinitrile ≥95.0%

Supplier: Decon Laboratories Ltd

U0059-25ML U0059-5ML
TCU0059-025MLEA 168 USD InStock
TCU0059-025ML TCU0059-5ML
Undecanedinitrile ≥95.0%
Undecanedinitrile
CAS Number: 71172-36-6
MDL Number: MFCD00039506
Molecular Formula: C11H18N2
Molecular Weight: 178.28
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 190
Specific Gravity (20/20): 0.91

Other Product Names / Synonyms: 1,9-Dicyanononane


Formula: C₁₁H₁₈N₂
Boiling Pt: 190 °C
Density: 0.91 (20 °C)
MDL Number: MFCD00039506
CAS Number: 71172-36-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f5de0a7800be71a430123ed09d89a743cfe2cb1&vid=8f5de0a7800be71a430123ed09d89a743cfe2cb1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty