Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Betanin (red beet extract diluted with dextrin)

Supplier: Decon Laboratories Ltd

B0397-25G B0397-100G
TCB0397-025GEA 88.84 USD InStock
TCB0397-025G TCB0397-100G
Betanin (red beet extract diluted with dextrin)
Betanin (red beet extract diluted with dextrin)
CAS Number: 7659-95-2
MDL Number: MFCD00060076
Form: Crystal
Color: Red

Product Categories: Bioscience, Glycosides, Alkaloids (Phytochemicals), Indole Alkaloids


Formula: C24H26N2O13
MW: 550.47 g/mol
MDL Number: MFCD00060076
CAS Number: 7659-95-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty