Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyltriethylammonium hydroxide 10% in water

Supplier: Decon Laboratories Ltd

B0445-25ML B0445-250ML
TCB0445-025MLEA 75 USD InStock
TCB0445-025ML TCB0445-250ML
Benzyltriethylammonium hydroxide 10% in water
Benzyltriethylammonium hydroxide
CAS Number: 1836-42-6
MDL Number: MFCD00040488
Molecular Formula: C13H23NO
Molecular Weight: 209.33
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.02


Formula: C₁₃H₂₃NO
Density: 1.02 (20 °C)
MDL Number: MFCD00040488
CAS Number: 1836-42-6
UN: 3286
ADR: 3,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e76ca0038e8a247e2437f875cc935eaedb6d177c&vid=e76ca0038e8a247e2437f875cc935eaedb6d177c&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty