Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitro-4-(trifluoromethoxy)aniline ≥97%

Supplier: MATRIX SCIENTIFIC MS

001349-25G 001349-5G 001349-100G
101775-352EA 75.38 USD InStock
101775-352 101775-350 101775-354
2-Nitro-4-(trifluoromethoxy)aniline ≥97%
2-Nitro-4-(trifluoromethoxy)aniline
Hazard: Toxic


Formula: C₇H₅F₃N₂O₃
Melting Pt: 62-64°C
MDL Number: MFCD00042326
CAS Number: 2267-23-4
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Melting point 62-64°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty