Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-chlorophenyl isothiocyanate ≥98%

Supplier: MATRIX SCIENTIFIC MS

001649-1G 001649-5G
101775-478EA 63.08 USD InStock
101775-478 101775-480
4-Bromo-2-chlorophenyl isothiocyanate ≥98%
4-Bromo-2-chlorophenyl isothiocyanate
Hazard: Toxic/Corrosive/Refrigerate/Lachrymator


Formula: C₇H₃BrClNS
Boiling Pt: 133…134 °C (0.5 mmHg)
Melting Pt: 42…44 °C
MDL Number: MFCD00041051
CAS Number: 98041-69-1
UN: 2923
ADR: 8,III
Order Now
Specification Test Results
Boiling point 133-134°/0

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty