Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Chloro-1-iodotetrafluoroethane ≥97%

Supplier: MATRIX SCIENTIFIC MS

001919-5G 001919-25G
101775-602EA 70.77 USD InStock
101775-602 101775-604
1-Chloro-1-iodotetrafluoroethane ≥97%
1-Chloro-1-iodotetrafluoroethane
Hazard: Toxic/Light Sensitive


Formula: C₂ClF₄I
Boiling Pt: 55…56 °C
CAS Number: 754-23-4
UN: 2810
ADR: 6.1,II
Order Now
Specification Test Results
Boiling point 55-56°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty