Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2,4-difluorobenzoyl chloride

Supplier: MATRIX SCIENTIFIC MS

001948-1G 001948-5G
101775-614EA 29.23 USD InStock
101775-614 101775-616
3-Chloro-2,4-difluorobenzoyl chloride
3-Chloro-2,4-difluorobenzoyl chloride
Hazard: Corrosive/Moisture Sensitive


Formula: C₇H₂Cl₂F₂O
MW: 210.99 g/mol
Boiling Pt: 222
Melting Pt: 70
CAS Number: 157373-00-7
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty