Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methyl-2-(trifluoromethyl)furo-3-ylacetonitrile

Supplier: MATRIX SCIENTIFIC MS

002000-250MG 002000-1G
101775-660EA 60 USD InStock
101775-660 101775-662
5-Methyl-2-(trifluoromethyl)furo-3-ylacetonitrile
5-Methyl-2-(trifluoromethyl)furo-3-ylacetonitrile
Hazard: Irritant


Formula: C₉H₆F₃NO₂
Melting Pt: 75…76 °C
MDL Number: MFCD00277690
CAS Number: 175276-72-9
Order Now
Specification Test Results
Melting point 75-76°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty