Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1,1,2,2-Tetrafluoroethoxy)toluene ≥98%

Supplier: MATRIX SCIENTIFIC MS

002036-5G 002036-25G
101775-696EA 38.46 USD InStock
101775-696 101775-698
2-(1,1,2,2-Tetrafluoroethoxy)toluene ≥98%
2-(1,1,2,2-Tetrafluoroethoxy)toluene
Hazard: Toxic


Formula: C₉H₈F₄O
Boiling Pt: 58…60 °C (14 mmHg)
Density: 1.26
MDL Number: MFCD00042103
CAS Number: 42145-66-4
Order Now
Specification Test Results
Boiling point 58-60°/14m
Density 1.26

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty