Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-fluorobenzoyl chloride

Supplier: MATRIX SCIENTIFIC MS

002094-5G 002094-25G
101775-758EA 60 USD InStock
101775-758 101775-760
3-Chloro-2-fluorobenzoyl chloride
3-Chloro-2-fluorobenzoyl chloride
Hazard: Corrosive


Formula: C₇H₃Cl₂FO
MW: 193
Boiling Pt: 202…203 °C
Density: 1.451
MDL Number: MFCD01631548
CAS Number: 85345-76-2
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Boiling point 202-203°
Density 1.451

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty