Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-2-methylbenzenesulphonyl chloride ≥97%

Supplier: MATRIX SCIENTIFIC MS

002176-5G 002176-25G
101775-832EA 26.15 USD InStock
101775-832 101775-834
5-Fluoro-2-methylbenzenesulphonyl chloride ≥97%
5-Fluoro-2-methylbenzenesulphonyl chloride
Hazard: Corrosive


Formula: C₇H₆ClFO₂S
Boiling Pt: 130…132 °C (15 mmHg)
Density: 1.43
MDL Number: MFCD00052958
CAS Number: 445-05-6
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Boiling point 130-132°/1

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty