Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H,1H,2H,2H-Perfluoro-1-iodooctane ≥95%

Supplier: MATRIX SCIENTIFIC MS

002227-100G 002227-25G
101775-878EA 112.31 USD InStock
101775-878 101775-876
1H,1H,2H,2H-Perfluoro-1-iodooctane ≥95%
1H,1H,2H,2H-Perfluoro-1-iodooctane
TCA listed: T
Hazard: Irritant-Harmful/Refrigerate/Light Sensitive


Formula: C₈H₄F₁₃I
MW: 474
Boiling Pt: 179
Melting Pt: 20.5
Density: 1.94
Flash Pt: 150 °C
MDL Number: MFCD00039410
CAS Number: 2043-57-4
Order Now
Specification Test Results
Boiling point 178-180°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty