Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-(trifluoromethyl)benzonitrile ≥97%

Supplier: MATRIX SCIENTIFIC MS

002274-1G 002274-5G
101775-916EA 21.54 USD InStock
101775-916 101775-918
2-Fluoro-3-(trifluoromethyl)benzonitrile ≥97%
2-Fluoro-3-(trifluoromethyl)benzonitrile
Hazard: Toxic


Formula: C₈H₃F₄N
MW: 189.11
Boiling Pt: 212
Melting Pt: 28
Density: 1.323
MDL Number: MFCD00061152
CAS Number: 146070-35-1
UN: 3276
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty