Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-(trifluoromethyl)pyrimidine-5-carbonyl chloride

Supplier: MATRIX SCIENTIFIC MS

002465-1G 002465-5G
101776-102EA 130.77 USD InStock
101776-102 101776-104
2-Chloro-4-(trifluoromethyl)pyrimidine-5-carbonyl chloride
2-Chloro-4-(trifluoromethyl)pyrimidine-5-carbonyl chloride
Hazard: Corrosive/Moisture Sensitive


Formula: C₆HCl₂F₃N₂O
Boiling Pt: 54 °C (0.15 mmHg)
MDL Number: MFCD00173789
CAS Number: 154934-99-3
Order Now
Specification Test Results
Boiling point 54°/0.15mm

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty