Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-fluoroaniline ≥98%

Supplier: MATRIX SCIENTIFIC MS

002490-100G 002490-25G
101776-124EA 75.38 USD InStock
101776-124 101776-122
2-Chloro-4-fluoroaniline ≥98%
2-Chloro-4-fluoroaniline
TCA listed: T
Hazard: Toxic


Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 192
Density: 1.219
MDL Number: MFCD00042530
CAS Number: 2106-02-7
UN: 2810
ADR: 6.1 III
Order Now
Specification Test Results
Boiling point 192°
Density 1.219
ND20 1.554

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty