Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-fluorobenzonitrile ≥99%

Supplier: MATRIX SCIENTIFIC MS

002498-25G 002498-100G
101776-130EA 56.92 USD InStock
101776-130 101776-132
3-Chloro-4-fluorobenzonitrile ≥99%
3-Chloro-4-fluorobenzonitrile
Hazard: Toxic/Irritant


Formula: C₇H₃ClFN
MW: 155.56
Boiling Pt: 135
Melting Pt: 67…69 °C
MDL Number: MFCD00015431
CAS Number: 117482-84-5
UN: 3439
ADR: 6.1 III
Order Now
Specification Test Results
Melting point 67-69°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty