Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-toluoyl chloride

Supplier: MATRIX SCIENTIFIC MS

002575-1G 002575-5G
101776-190EA 24.62 USD InStock
101776-190 101776-192
3-Fluoro-4-toluoyl chloride
3-Fluoro-4-toluoyl chloride
Hazard: Corrosive


Formula: C₈H₆ClFO
MW: 172.59 g/mol
Boiling Pt: 215
Melting Pt: 38
MDL Number: MFCD00671774
CAS Number: 59189-97-8
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty