Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-p-cresol ≥98%

Supplier: MATRIX SCIENTIFIC MS

002585-25G 002585-5G
101776-208EA 100 USD InStock
101776-208 101776-206
3-Chloro-p-cresol ≥98%
3-Chloro-p-cresol
Hazard: Corrosive/Irritant


Formula: C₇H₇ClO
Boiling Pt: 228 °C
Melting Pt: 55…56 °C
MDL Number: MFCD00060319
CAS Number: 615-62-3
UN: 3437
ADR: 6.1,II
Order Now
Specification Test Results
Boiling point 228°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty