Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-(trifluoromethyl)benzyl bromide

Supplier: MATRIX SCIENTIFIC MS

002642-1G 002642-5G 002642-25G
101776-246EA 38.46 USD InStock
101776-246 101776-248 101776-250
2-Chloro-5-(trifluoromethyl)benzyl bromide
2-Chloro-5-(trifluoromethyl)benzyl bromide
Hazard: Corrosive


Formula: C₈H₅BrClF₃
MW: 273.48
Boiling Pt: 232
Melting Pt: 51
MDL Number: MFCD00236216
CAS Number: 237761-77-2
UN: 3265
ADR: 8 II
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty