Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-(trifluoromethyl)benzoyl chloride ≥97%

Supplier: MATRIX SCIENTIFIC MS

002671-25G 002671-5G
101776-276EA 184.62 USD InStock
101776-276 101776-274
3-Fluoro-5-(trifluoromethyl)benzoyl chloride ≥97%
3-Fluoro-5-(trifluoromethyl)benzoyl chloride
Hazard: Corrosive/Moisture Sensitive


Formula: C₈H₃ClF₄O
MW: 226.56
Boiling Pt: 119
Density: 1.482
MDL Number: MFCD00061157
CAS Number: 171243-30-4
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Boiling point 119°
Density 1.482
ND20 1.462

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty