Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-fluorobenzylamine ≥98%

Supplier: MATRIX SCIENTIFIC MS

002775-5G 002775-25G
101776-370EA 46.15 USD InStock
101776-370 101776-372
2-Chloro-6-fluorobenzylamine ≥98%
2-Chloro-6-fluorobenzylamine
Hazard: Corrosive


Formula: C₇H₇ClFN
Boiling Pt: 91-93°C/15mm
MDL Number: MFCD00042458
CAS Number: 15205-15-9
Order Now
Specification Test Results
Boiling point 91-93°/15m

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty