Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromopentafluoroacetone ≥98%

Supplier: MATRIX SCIENTIFIC MS

002943-5G 002943-25G
101776-444EA 438.46 USD InStock
101776-444 101776-446
Bromopentafluoroacetone ≥98%
Bromopentafluoroacetone
Hazard: Corrosive/Irritant/Light Sensitive/Refrigerate


Formula: C₃BrF₅O
Boiling Pt: 31…33 °C
MDL Number: MFCD00153080
CAS Number: 815-23-6
Order Now
Specification Test Results
Boiling point 31-33°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty