Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-isothiocyanatobenzoate ≥98%

Supplier: MATRIX SCIENTIFIC MS

003061-1G 003061-5G
101776-486EA 67.69 USD InStock
101776-486 101776-488
Methyl-4-isothiocyanatobenzoate ≥98%
Methyl 4-isothiocyanatobenzoate
Hazard: Toxic/Corrosive/Refrigerate/Lachrymator


Formula: C₉H₇NO₂S
Melting Pt: 46-48°C
MDL Number: MFCD00041099
CAS Number: 3662-78-0
Order Now
Specification Test Results
Melting point 46-48°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty