Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α,α-Trifluoro-m-xylene

Supplier: MATRIX SCIENTIFIC MS

003310-1G 003310-5G
101776-606EA 21.54 USD InStock
101776-606 101776-608
α,α,α-Trifluoro-m-xylene
α,α,α-Trifluoro-m-xylene
Hazard: Flammable


Formula: C₈H₇F₃
Boiling Pt: 130°C
Density: 1.148
Flash Pt: 26°C
MDL Number: MFCD00190105
CAS Number: 401-79-6
Order Now
Specification Test Results
Boiling point 130°
Density 1.148
ND20 1.427

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty