Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethoxybenzoyl chloride ≥98%

Supplier: MATRIX SCIENTIFIC MS

003314-25G 003314-5G
101776-616EA 154.31 USD InStock
101776-616 101776-614
4-Ethoxybenzoyl chloride ≥98%
4-Ethoxybenzoyl chloride
Hazard: Corrosive/Lachrymator


Formula: C₉H₉ClO₂
Boiling Pt: 142-145°C/20mm
Density: 1.2
MDL Number: MFCD00000688
CAS Number: 16331-46-7
Order Now
Specification Test Results
Boiling point 142-145°/2
Density 1.2
ND20 1.5672

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty