Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chlorotetrafluoropropionyl chloride ≥97%

Supplier: MATRIX SCIENTIFIC MS

003496-1G 003496-5G
101776-764EA 1256.92 USD InStock
101776-764 101776-766
3-Chlorotetrafluoropropionyl chloride ≥97%
3-Chlorotetrafluoropropionyl chloride
Hazard: Corrosive/Refrigerate


Formula: C₃Cl₂F₄O
MW: 198.93 g/mol
Boiling Pt: 49…51 °C
MDL Number: MFCD00153100
CAS Number: 24503-62-6
Order Now
Specification Test Results
Boiling point 49-51°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty