Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,3,3-Tetrafluoropropyl methacrylate ≥97%

Supplier: MATRIX SCIENTIFIC MS

003501-100G 003501-25G
101776-774EA 212.31 USD InStock
101776-774 101776-772
2,2,3,3-Tetrafluoropropyl methacrylate ≥97%
2,2,3,3-Tetrafluoropropyl methacrylate
Hazard: Flammable


Formula: C₇H₈F₄O₂
Boiling Pt: 70 °C (51 mmHg)
Density: 1.254
MDL Number: MFCD00042381
CAS Number: 45102-52-1
Order Now
Specification Test Results
Boiling point 70°/51mm
Density 1.254
ND20 1.374

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty