Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3,4,4-Tetrafluorotetrahydrofuran ≥98%

Supplier: MATRIX SCIENTIFIC MS

003504-5G 003504-1G
101776-778EA 946.15 USD InStock
101776-778 101776-776
3,3,4,4-Tetrafluorotetrahydrofuran ≥98%
3,3,4,4-Tetrafluorotetrahydrofuran
Hazard: Flammable


Formula: C₄H₄F₄O
Boiling Pt: 82 °C
MDL Number: MFCD02093328
CAS Number: 374-33-4
Order Now
Specification Test Results
Boiling point 82°

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty