Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,1-Trifluoro-2,3-epoxypropane ≥98%

Supplier: MATRIX SCIENTIFIC MS

003563-25G 003563-5G
101776-820EA 433.85 USD InStock
101776-820 101776-818
1,1,1-Trifluoro-2,3-epoxypropane ≥98%
1,1,1-Trifluoro-2,3-epoxypropane
Hazard: Flammable/Toxic/Stored under Argon/Moisture Sensitive/Refrigerate


Formula: C₃H₃F₃O
Boiling Pt: 38-40°C
Density: 1.3068
Flash Pt: -40°C
MDL Number: MFCD00041506
CAS Number: 359-41-1
Order Now
Specification Test Results
Boiling point 38-40°
Density 1.3068
ND20 1.2997

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty