Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl trifluoromethanesulphonate ≥98%

Supplier: MATRIX SCIENTIFIC MS

003753-5G 003753-25G
101777-094EA 36.92 USD InStock
101777-094 101777-096
Ethyl trifluoromethanesulphonate ≥98%
Ethyl trifluoromethanesulphonate
TCA listed: T
Hazard: Flammable/Toxic


Formula: C₃H₅F₃O₃S
MW: 178.13 g/mol
Boiling Pt: 115 °C
Density: 1.374
MDL Number: MFCD00000410
CAS Number: 425-75-2
UN: 2920
ADR: 8 II
Order Now
Specification Test Results
Boiling point 115°
Density 1.374
ND20 1.3356

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty