Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chlorothioanisole ≥99%

Supplier: MATRIX SCIENTIFIC MS

003952-5G 003952-25G
101777-252EA 24.62 USD InStock
101777-252 101777-254
3-Chlorothioanisole ≥99%
3-Chlorothioanisole
Hazard: Irritant/Stench


Formula: C₇H₇ClS
MW: 158.65 g/mol
Boiling Pt: 44
Density: 1.206
Flash Pt: 88 °C
MDL Number: MFCD00040127
CAS Number: 4867-37-2
UN: 3334
ADR: 9
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty