Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetylthiophene ≥99%

Supplier: MATRIX SCIENTIFIC MS

003962-500G 003962-100G
101777-270EA 141.54 USD InStock
101777-270 101777-268
2-Acetylthiophene ≥99%
2-Acetylthiophene
TCA listed: T
Hazard: Toxic/Stench


Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 214
Melting Pt: 9
MDL Number: MFCD00005442
CAS Number: 88-15-3
UN: 2810
ADR: 6.1 II
Order Now
Specification Test Results
Boiling point 78-80°/12m

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty