Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dimethylbenzylamine

Supplier: MATRIX SCIENTIFIC MS

004076-1G 004076-5G
101777-316EA 29.23 USD InStock
101777-316 101777-318
3,4-Dimethylbenzylamine
3,4-Dimethylbenzylamine
TCA listed: T
Hazard: Corrosive/Air Sensitive


Formula: C₉H₁₃N
Boiling Pt: 93°C/10mm
Density: 0.95
MDL Number: MFCD00025577
CAS Number: 102-48-7
Order Now
Specification Test Results
Boiling point 93°/10mm
Density 0.95

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty