Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclobutyl phenyl ketone ≥98%

Supplier: MATRIX SCIENTIFIC MS

004263-1G 004263-5G
101777-462EA 41.54 USD InStock
101777-462 101777-464
Cyclobutyl phenyl ketone ≥98%
Cyclobutyl phenyl ketone
Hazard: Irritant


Formula: C₁₁H₁₂O
MW: 160.2124 g/mol
Boiling Pt: 114…118 °C (7 mmHg)
Density: 1.05
MDL Number: MFCD00001326
CAS Number: 5407-98-7
Order Now
Specification Test Results
Boiling point 114-118°/7
Density 1.05
ND20 1.547

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty