Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl bromofluoroacetate ≥97%

Supplier: MATRIX SCIENTIFIC MS

004578-25G 004578-100G
101777-640EA 64.62 USD InStock
101777-640 101777-642
Ethyl bromofluoroacetate ≥97%
Ethyl bromofluoroacetate
TCA listed: T
Hazard: Toxic/Irritant/Lachrymator


Formula: BrFCHCOOCH₂CH₃
MW: 184.99 g/mol
Boiling Pt: 154
Density: 1.5407
MDL Number: MFCD00042095
CAS Number: 401-55-8
UN: 2810
ADR: 6.1 III
Order Now
Specification Test Results
Boiling point 154°
Density 1.5407
ND20 1.425

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty