Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoroanisole ≥98%

Supplier: MATRIX SCIENTIFIC MS

004645-100G 004645-500G 004645-1KG
101777-668EA 30.77 USD InStock
101777-668 101777-670 101777-672
2-Fluoroanisole ≥98%
2-Fluoroanisole
Hazard: Flammable


Formula: C₇H₇FO
Boiling Pt: 154-155°C
Melting Pt: -39°C
Density: 1.124
MDL Number: MFCD00000316
CAS Number: 321-28-8
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Boiling point 154-155°
Density 1.124
ND20 1.494

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty