Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexafluoroglutaric acid ≥98%

Supplier: MATRIX SCIENTIFIC MS

004878-25G 004878-100G
101777-804EA 118.46 USD InStock
101777-804 101777-806
Hexafluoroglutaric acid ≥98%
Hexafluoroglutaric acid
TCA listed: T
Hazard: Corrosive/Irritant/Hygroscopic


Formula: C₅H₂F₆O₄
Boiling Pt: 134-138°C/3mm
Melting Pt: 92-97°C
MDL Number: MFCD00004172
CAS Number: 376-73-8
UN: 3261
ADR: 8,III
Order Now
Specification Test Results
Boiling point 134-138°/3

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty