Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexafluoroglutaric anhydride ≥97%

Supplier: MATRIX SCIENTIFIC MS

004879-5G 004879-100G 004879-25G
101777-808EA 26.15 USD InStock
101777-808 101777-812 101777-810
Hexafluoroglutaric anhydride ≥97%
Hexafluoroglutaric anhydride
Hazard: Corrosive/Irritant/Moisture Sensitive


Formula: C₅F₆O₃
Boiling Pt: 72 °C
Density: 1.654
MDL Number: MFCD00006680
CAS Number: 376-68-1
UN: 3265
ADR: 8,III
Order Now
Specification Test Results
Boiling point 72°
Density 1.654
ND20 1.324

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty