Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Trifluoromethoxy)benzene ≥99%

Supplier: MATRIX SCIENTIFIC MS

004897-100G 004897-500G
101777-834EA 43.08 USD InStock
101777-834 101777-836
(Trifluoromethoxy)benzene ≥99%
(Trifluoromethoxy)benzene
TCA listed: T
Hazard: Flammable/Corrosive


Formula: C₇H₅F₃O
Boiling Pt: 102°C
Density: 1.226
MDL Number: MFCD00040832
CAS Number: 456-55-3
UN: 1993
ADR: 3,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=faf76f9047ed23a3eb2a61f855972f76a124b68f&vid=faf76f9047ed23a3eb2a61f855972f76a124b68f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Boiling point 102°
Density 1.226
ND20 1.406

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty